Learn More
Selleck Chemical LLC Periplocin S9181-5MG
SDP
Supplier:  Selleck Chemical LLC S91815MG
Encompass_Preferred
Periplocin (Periplocoside) extracted from the traditional herbal medicine cortex periplocae has cardiac and anticancer activity Periplocin could significantly boost proliferation migration and stimulate collagen production in fibroblast L929 cells which is dependent on activation of Src/ERK and PI3K/Akt pathways mediated by Na/K-ATPase and thus promoting wound healing
Catalog No. 50-000-24512