Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex High-Pressure Peristaltic Pump, Mid-Flow Range, 0.15 to 15 GPH, 125 PSI; 220 VAC
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7420428
Encompass
Choose a pump to achieve flow rates up to 158.5 GPH (598 LPH) and pressures up to 125 psi (8.6 bar) depending on your particular application. The pump performance allows for 30 feet (9 meters) of suction lift using water. Keeping chemical compatibility as
Catalog No. NC1754987