Learn More
Agilent Technologies PERISTALTIC PUMP TUBING,PK/12
Supplier:  Agilent Technologies 50050020
Encompass
Peristaltic pump tubing, Easy-Fit, standard for sample uptake, recommended for aqueous, acid or alkaline matrix, 1 pack of 12 pcs
Catalog No. NC2013704