Learn More
Medchemexpress LLC PERK-IN-4 | 1337531-89-1 | 98.21% | 469.43 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13781310MM/1ML
Encompass_Preferred
PERK-IN-4 is a potent and selective inhibitor of PERK (protein kinase R (PKR)-like endoplasmic reticulum kinase). It demonstrates an IC₅₀ of 0.3 nM. PERK is activated in response to various endoplasmic reticulum stresses, which are implicated in numerous disease states.
  • Potent and selective PERK inhibitor
  • IC₅₀ of 0.3 nM
  • Intended for research use in endoplasmic reticulum stress studies
Catalog No. 50-003-52193