Learn More
Medchemexpress LLC PERK-IN-6 | 1337532-14-5 | C23H22N6O | 5 MG

Supplier: Medchemexpress LLC HY1531605MG
PERK-IN-6 is a potent PERK inhibitor with an IC50 of 2.5 nM. It effectively inhibits thapsigargin-induced PERK autophosphorylation in A549 cells, showing an IC50 of 0.1-0.3 μM. This compound is intended for research use only.
- IC50 of 2.5 nM against PERK
- Inhibits thapsigargin induced PERK autophosphorylation
- Purity of 99.46%
- Appearance as an off-white to light yellow solid
- Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.