Learn More
McKesson General Medical Ethicon Perma-Hand Silk Suture, Non-Absorbable, Sterile, 30 in
Supplier:  McKesson General Medical 2891
Encompass
Ethicon PERMA-HAND Silk Suture is a non-absorbable, sterile braided suture made from silk, ideal for soft tissue approximation and ligation in various surgical procedures. It features a C-1 taper point needle for precise tissue penetration and a black color for visibility. Enhanced with a wax or silicone coating, it offers excellent handling and secure knot tying, making it a preferred choice among surgeons for reliable performance.
Catalog No. NC2879169