Learn More
Newcomer Supply PERMANENT MOUNTNG MEDIUM 125ML
SDP
Supplier:  Newcomer Supply NB300A
Encompass
Permanent Mounting medium preserves AEC DAB Fast Red and other dye chemistry stains. 125mlSets in 5 minutes at Room Temperature. Manufactured by Innovex Biosciences.
Catalog No. NC2311505