Learn More
Medchemexpress LLC Permanent Orange | 3468-63-1 | ≥97.0% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYD0352
Encompass_Preferred
Permanent Orange is a synthetic pigment commonly used in the manufacture of paints and coatings. It is an organic pigment composed of carbon and other elements, delivering a vibrant orange color. This pigment boasts fade and weather resistance, making it ideal for outdoor applications. It can be combined with other pigments to achieve a diverse palette of colors and is compatible with various binders and solvents. Additionally, it serves as a biochemical reagent suitable for life science-related research.
  • Synthetic organic pigment
  • Vibrant orange color
  • Fade and weather resistant
  • Ideal for outdoor applications
  • Compatible with various binders and solvents
  • Suitable as a biochemical reagent for life science research
Catalog No. 50-004-13935