Learn More
Electron Microscopy Sciences Lab-Tek 4 Chambers Slide, Permanox, 2 trays
SDP
Supplier:  Electron Microscopy Sciences 70400
Encompass_Preferred
Chamber slides combine a media-containing chambered super-structure with a standard soda lime glass slide or Permanox plastic slide, sealing the two together with a biologically inert silicone gasket. After cells are grown, the chamber and gasket are easily removed. The slide may then be fixed, stained, cover slipped, and filed with the usual convenience. These slides, once fixed and stained, can be embedded.
Packed in a sterile sealed tray, 8 units per tray. Permanox does not interfere with fluorescent label.
Material: Permanox
Package: 2 trays
Catalog No. 50-189-9293