Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bellco Glass, Inc. Permeable Cell Culture Inserts, Packed in 6 Well Plate, PC, 8.0µm, Sterile
SDP

Supplier:  Bellco Glass, Inc. 230605000

Encompass_Preferred

Permeable Cell Culture Inserts mimic in vivo conditions. Inserts center in wells, preventing membrane from touching sides and wicking between insert and side wall. Membrane remains suspended above well bottom, allowing co-culture: grow one cell line inside insert, and another below in the well. Ideal for drug transfer assays, transport studies, migration assays, and cell-cell interaction studies. Non-Treated Plates sold separately; carrier plate is Tissue Culture Treated which encourages cell adhesion.\n\nSpecifications:\n

    \n
  • 0.4µm pore size is ideal for co-culture and drug transport studies
  • \n
  • 3.0µm and 8.0µm pore size are ideal for angiogenesis and chemotaxis experiments
  • \n
  • Membrane Material: PC
  • \n
  • Membrane Pore Size: 0.4um
  • \n
  • Case Quantity: 12
  • \n

Catalog No. 50-233-0232


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.