Learn More
Medchemexpress LLC Perospirone hydrochloride | 129273-38-7 | 99.9% | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYB073110MM/1ML
Encompass_Preferred
Perospirone hydrochloride (SM-9018) is an orally active antagonist of the 5-HT2A receptor (Ki of 0.6 nM) and dopamine D2 receptor (Ki of 1.4 nM). It also acts as a partial agonist of the 5-HT1A receptor (Ki of 2.9 nM). This compound is classified as an atypical antipsychotic agent and has potential for research related to schizophrenic disease. It is for research use only and not sold to patients.
  • Orally active antagonist of the 5-HT2A receptor (Ki of 0.6 nM)
  • Antagonist of the dopamine D2 receptor (Ki of 1.4 nM)
  • Partial agonist of the 5-HT1A receptor (Ki of 2.9 nM)
  • Classified as an atypical antipsychotic agent
  • Potential for research into schizophrenic disease
Catalog No. 50-004-08190