Learn More
Medchemexpress LLC Perospirone (hydrochloride) | 129273-38-7 | 99.9% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYB0731
Encompass_Preferred
Perospirone hydrochloride (SM-9018) is an orally active antagonist of the 5-HT2A receptor (Ki of 0.6 nM) and dopamine D2 receptor (Ki of 1.4 nM). It also acts as a partial agonist of the 5-HT1A receptor (Ki of 2.9 nM). This compound is classified as an atypical antipsychotic agent and is being investigated for its potential in schizophrenic disease research.
  • Orally active 5-HT2A receptor antagonist (Ki of 0.6 nM)
  • Dopamine D2 receptor antagonist (Ki of 1.4 nM)
  • Partial agonist of the 5-HT1A receptor (Ki of 2.9 nM)
  • Atypical antipsychotic agent
  • Potential for schizophrenic disease research
  • Targets 5-HT1 Receptor (Ki: 18 nM)
  • Targets D1 Receptor (Ki: 41 nM)
Catalog No. 50-004-50966