Learn More
Diagnostic Biosystems PEROXIDASE BLOCK Peroxidase Block
Supplier:  Diagnostic Biosystems K033
Encompass_Preferred
This product is designed for use in peroxidase based immunohistochemical staining procedures done on paraffin tissue sections to suppress nonspecific staining due to endogenous peroxidase activity.\n\nEndogenous peroxidase activity has been observed in tissue sections during immunoperoxidase staining. If not properly suppressed, endogenous peroxidase can produce false signal as well as high background during the immunostaining. Treatment of tissue sections with Peroxidase Block prior to the application of primary antibody will suppress endogenous peroxidase activity.
Catalog No. NC2049159