Learn More
Medchemexpress LLC Peroxiredoxin 1 Antibody (YA692) | 10 UL

Supplier: Medchemexpress LLC HYP8078210UL
This Peroxiredoxin 1 Antibody (YA692) is a mouse-derived antibody exhibiting reactivity with human, mouse, and rat samples. It has a molecular weight of 22 kDa and is supplied at a concentration of 1.72 mg/mL. This antibody is suitable for various applications including Western Blot (WB), Immunohistochemistry (frozen and paraffin-embedded sections), and Immunocytochemistry/Immunofluorescence (ICC/IF).
- Host: Mouse
- Reactivity with human, mouse, and rat samples
- Predicted and observed molecular weight: 22 kDa
- Concentration: 1.72 mg/mL
- Validated for Western Blot (WB), IHC-F, IHC-P, and ICC/IF applications
- Formulated in 1*PBS (pH 7.3), 50% glycerol, and 0.5% BSA
- Contains 0.02% sodium azide as a preservative
- Stable for 1 year when stored at -20°C
- Avoid repeated freeze/thaw cycles to maintain integrity
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.