Learn More
Medchemexpress LLC Peroxiredoxin 5 antibody (YA1591)(PBS only) | 50 UL

Supplier: Medchemexpress LLC HYP8184650UL
The Peroxiredoxin 5 Antibody (YA1591) is a rabbit-derived, non-conjugated IgG monoclonal antibody targeting Peroxiredoxin 5. It reacts with human, mouse, and rat samples. This antibody is crucial for understanding the antioxidant enzyme family, which reduces hydrogen peroxide and alkyl hydroperoxides, interacting with peroxisome receptor 1 for tissue protection.
- Rabbit-derived IgG monoclonal antibody
- Targets Peroxiredoxin 5
- Reacts with human, mouse, and rat
- Applications: WB, IHC-P, IP
- Provided in PBS, pH 7.4
- Affinity purified
- Store at -20°C for 1 year
- Avoid freeze/thaw cycles
- Predicted band size: 22 kDa; Observed: 17 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.