Learn More
Medchemexpress LLC HY-A0077 500mg Medchemexpress, Perphenazine CAS:58-39-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0077500MG
Encompass_Preferred
Medchemexpress, HY-A0077 500mg Perphenazine CAS:58-39-9 Perphenazine is a typical antipsychotic drug, inhibits 5-HT2Areceptor, Alpha-1A adrenergic receptor, Dopamine receptor D2/D3, D2L receptor, and Histamine H1 receptor, with Ki values of 5.6, 10, 0.765/0.13, 3.4, and 8 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1001