Learn More
Medchemexpress LLC Perphenazine-d4 | 155593-75-2 | 98.8% | C21H22D4ClN3OS | 5 MG
SDP
Supplier:  Medchemexpress LLC HYA0077S15MG
Encompass_Preferred
Perphenazine-d4 is the deuterium labeled Perphenazine, a typical antipsychotic agent. It inhibits several receptors including 5-HT2A, Alpha-1A adrenergic, Dopamine D2/D3, D2L, and Histamine H1 receptors. Deuteration can affect the pharmacokinetic and metabolic profiles of drugs, making Perphenazine-d4 useful as a tracer for quantitation in drug development.
  • Deuterium labeled Perphenazine
  • Typical antipsychotic agent
  • Inhibits 5-HT2A receptor, Alpha-1A adrenergic receptor, Dopamine receptor D2/D3, D2L receptor, and Histamine H1 receptor
  • Useful as tracers for quantitation during drug development
  • Relevant in studies related to 5-HT Receptor, Adrenergic Receptor, Apoptosis, Autophagy, Dopamine Receptor, and Histamine Receptor pathways
  • Applicable in research involving GPCR/G Protein, Neuronal Signaling, Apoptosis, Autophagy, and Immunology/Inflammation pathways
Catalog No. 50-004-32773