Learn More
Medchemexpress LLC Pertuzumab, 10mg | CAS No. : 380610-27-5
SDP
Supplier:  Medchemexpress LLC HYP991210MG
Encompass_Preferred
Pertuzumab, a humanized IgG1 monoclonal antibody, is a HER2 dimerization inhibitor for the treatment of metastatic HER2-positive breast cancer.
Catalog No. NC3823048