Learn More
Uline Polyester Strapping - 1/2" x .028" x 6, 500', Black
Supplier:  Uline S12530
Encompass_Preferred
Replaces steel strapping for medium and heavy-duty applications. 70% less expensive than steel. 80% lighter, easier to use. Less elongation than poly, more flexibility than steel.
Catalog No. NC2022659