Learn More
Sterlitech Corporation Polyester (PETE) Membrane Filters, Transparent, 3.0 Micron, 12 Micron Thickness, 6E5 pores/cm2, 90mm, 30/pack
SDP
Supplier:  Sterlitech Corporation 1300028
Encompass_Preferred
Sterlitech solvent resistant, hydrophilic polyester track etch (PETE) membranes are made from a thin, translucent, microporous, polyester film and are ideal for use in blood assays, microscopic analysis, and the general filtration of samples.PETE Membrane Filter Applications:* Precise General Filtration and Prefiltration* Removal of red blood cells from plasma* Flow control of reagents through assay
Catalog No. 50-195-2352