Learn More
Ted Pella Inc PETRI DISH 100/PKG
SDP
Supplier:  Ted Pella Inc 36135
Encompass
Polypropylene Petri Dishes, Suitable for Use in the Microwave Systems, Bottom Dimensions: 55mm O.D., 53mm I.D., Top Dimensions: 58mm O.D., 56mm I.D., Total Assembled Height: 15mm
Catalog No. NC9881254