Learn More
Bellco Glass, Inc. Petri Dish Spreader,90・
SDP
Supplier:  Bellco Glass, Inc. 565400090
Encompass_Preferred
Ideal for spreading Inoculum over agar face in Petri dish cultures. Manufactured from Type 1 Class A Borosilicate Glass Rod. MADE IN THE USA Length: 130mm Style bend: 90 degrees
Catalog No. 50-194-5834