Learn More
Nelson Jameson Company 3M Petrifilm Rapid Yeast and Mold Count Plate, Pack Size: 25 plates per foil pouch (50 per box), 500 plates per case, Incubation Time: 48-72 hours, Storage: -20 to 8°C
Supplier:  Nelson Jameson Company 4603477 C
Encompass
The 3M™ Petrifilm™ Rapid Yeast and Mold Count Plate is a efficient culture medium system designed for rapid and reliable enumeration of yeasts and molds in various samples including food, beverage, and environmental products. This innovative solution reduces incubation time compared to traditional methods and brings reliable results within 48 hours, enhancing laboratory productivity and workflow.
  • Fast results in 48 hours
  • Eliminates need for agar preparation
  • Streamlined laboratory workflow
  • Compatible with various diluents
  • Up to 150 colonies can be counted per plate
  • Storage at -20 to 8°C
  • Colony appearance includes distinct characteristics for yeast and mold
Catalog No. NC2745331