Learn More
Cayman Chemical Petromyzonl 10mg
SDP
Supplier:  Cayman Chemical 9825010 MG
Encompass_Preferred
Petromyzonol is a tetrahydroxy stearol which serves as the primary spawning pheromone in adult sea lamprey. It is produced in the bile of sea lamprey larvae from the bile acid precursor allocholic acid. While the adult sea lamprey is relatively insensitive to petromyzonol itself, the C-24 sulfate ester (petromyzonol sulfate) is a pheromone and a spawning chemoattractant which can be detected at very low concentrations by lamprey olfactory chemoreceptors. Petromyzonol, petromyzonol sulfate, and allocholic acid are all found in water samples from fresh water streams bearing larval lamprey populations.{7478}
Catalog No. 50-289-5011