Learn More
Medchemexpress LLC Petroselaidic acid | 593-40-8 | MFCD00069607 | 98.0% | 282.46 g/mol | C18H34O2 | 10 MG

Supplier: Medchemexpress LLC HYW34297910MG
Petroselaidic acid is a trans monounsaturated fatty acid and the trans isomer of petroselinic acid. It occurs in dairy products and is supplied as a research-grade solid for analytical, biochemical, and standards use.
- High purity: 98.0%.
- Chemical formula C18H34O2; molecular weight 282.46 g/mol.
- Physical form: solid powder.
- Storage: powder -20°C, 3 years; in solvent -80°C for 6 months or -20°C for 1 month.
- Applications: analytical and biochemical research, reference standards.
- Packaging: available in small quantities (e.g., 5 mg, 10 mg); larger sizes by quote.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.