Learn More
Medchemexpress LLC (+)-Peusedanol | 20516-23-8 | 5 MG

Supplier: Medchemexpress LLC HYN60635MG
(+)-Peusedanol is a coumarin isolated from *Peucedanum japonicum*. This white to off-white solid has a molecular formula of C14H16O5 and a molecular weight of 264.27. It is typically used for research purposes.
- High purity: 99.83%
- Soluble in ethanol at 12.5 mg/mL
- Store at 4°C, sealed, away from moisture and light
- If in solvent, store at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.