Learn More
Medchemexpress LLC (+)-Peusedanol | 20516-23-8 | 99.8% | 264.27 g/mol | C14H16O5 | 10 MG

Supplier: Medchemexpress LLC HYN606310MG
(+)-Peusedanol is a naturally occurring coumarin isolated from Peucedanum japonicum, supplied for research and analytical applications. The compound is provided as a high-purity solid with supporting COA and SDS documentation available for laboratory use.
- High purity (99.83%).
- Molecular formula C14H16O5.
- Molecular weight 264.27 g/mol.
- Supplied in small quantities suitable for analytical work.
- Certificate of analysis and safety data sheet available for download.
- Store sealed, away from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.