Learn More
Medchemexpress LLC HY-70062 10mg Medchemexpress, Pevonedistat CAS:905579-51-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY7006210MG
Encompass_Preferred
Medchemexpress, HY-70062 10mg Pevonedistat CAS:905579-51-3 Pevonedistat (MLN4924) is a potent and selective NEDD8-activating enzyme (NAE) inhibitor with an IC50 of 4.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9362
 Restricted Item