Learn More
Selleck Chemical LLC Pevonedistat
SDP
Supplier:  Selleck Chemical LLC S710925MG
Encompass_Preferred
MLN4924 is a small molecule inhibitor of Nedd8 activating enzyme (NAE) with IC50 of 4 nM
Catalog No. 50-000-00060