Learn More
Medchemexpress LLC HY-10484 1mg Medchemexpress, Pevonedistat hydrochloride CAS:1160295-21-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY104841MG
Encompass_Preferred
Medchemexpress, HY-10484 1mg Pevonedistat HCl CAS:1160295-21-5 Pevonedistat hydrochloride (MLN4924 hydrochloride) is a potent and selective NEDD8-activating enzyme (NAE) inhibitor, with an IC50 of 4.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8234