Learn More
MSC Pewag Oblong Master Link, G100, Alloy Steel, 0.75 in Diameter
Supplier: MSC 60470713
This master link is constructed from G100 grade alloy steel, offering durability and strength for various lifting applications. Its oblong shape and robust design are engineered to provide reliable performance with a significant work load limit.
- Oblong shape for versatile attachment
- G100 material grade ensures high strength
- Alloy steel construction for enhanced durability
- 0.75 in overall diameter
- Work load limit of 12800 lb