Learn More
Cayman Chemical PexIdartInIb 10mg
SDP
Supplier:  Cayman Chemical 1827110 MG
Encompass_Preferred
A brain-penetrant inhibitor of CSF1R, as well as c-Kit and FLT3 (IC50s = 20, 10, and 160 nM in vitro, respectively); blocks macrophage recruitment in mammary tumor-bearing mice and blocks glioblastoma invasion in both cell culture and a mouse model of glioblastoma multiforme
Catalog No. 50-298-0108