Learn More
Selleck Chemical LLC Pexidartinib
SDP
Supplier:  Selleck Chemical LLC S7818200MG
Encompass_Preferred
Pexidartinib (PLX3397) is an oral potent multi-targeted receptor tyrosine kinase inhibitor of CSF-1R Kit (c-Kit) and FLT3 with IC50 of 20 nM 10 nM and 160 nM respectively Pexidartinib (PLX3397) induces apoptosis and necrosis with antitumor activity Phase 3
Catalog No. 50-000-00055