Learn More
Medchemexpress LLC HY-16749A 50mg Medchemexpress, Pexidartinib (hydrochloride) CAS:2040295-03-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY16749A50MG
Encompass_Preferred
Medchemexpress, HY-16749A 50mg Pexidartinib (hydrochloride) CAS:2040295-03-0 Pexidartinib hydrochloride (PLX-3397 hydrochloride) is a potent, orally active, selective, and ATP-competitive colony stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit inhibitor, with IC50s of 20 and 10 nM, respectively. Pexidartinib hydrochloride exhibits 10- to 100-fold selectivity for c-Kit and CSF1R over other related kinases. Pexidartinib hydrochloride induces cell apoptosis and has anti-cancer activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9217