Learn More
Medchemexpress LLC Pexidartinib hydrochloride | 2040295-03-0 | 454.28 g/mol | C20H16Cl2F3N5 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY16749A10MG
Encompass_Preferred
Pexidartinib hydrochloride is a research-grade small-molecule tyrosine kinase inhibitor that selectively targets colony stimulating factor 1 receptor (CSF1R) and c-Kit. It is used in preclinical studies to investigate signaling pathways, tumor biology, and kinase-driven cellular responses. The compound is provided as the hydrochloride salt and is intended for laboratory research use only.
  • Selective CSF1R and c-Kit inhibitor with low-nanomolar potency.
  • Provided as the hydrochloride salt for stability and handling.
  • Suitable for in vitro and in vivo preclinical research; not for human use.
  • Characterized by CAS number 2040295-03-0 for unambiguous identification.
  • Molecular formula C20H16Cl2F3N5; molecular weight 454.28 g/mol.
  • Available in small pack sizes appropriate for laboratory experiments, including 10 mg.
Catalog No. 50-002-89644