Learn More
Medchemexpress LLC Pexidartinib hydrochloride | 2040295-03-0 | 99.8% | 454.28 | 2 G
SDP
Supplier:  Medchemexpress LLC HY16749A2G
Encompass_Preferred
Pexidartinib hydrochloride is a potent, orally active, selective, and ATP-competitive inhibitor of colony-stimulating factor 1 receptor (CSF1R or M-CSFR) and c-Kit. It exhibits 10- to 100-fold selectivity for c-Kit and CSF1R over other related kinases and induces cell apoptosis, showing anti-cancer activity.
  • Potent and selective inhibition of CSF1R and c-Kit.
  • Induces cell apoptosis.
  • Demonstrates anti-cancer activity.
  • Shows 10- to 100-fold selectivity over other related kinases.
Catalog No. 50-003-91148