Learn More
Medchemexpress LLC Pexopiprant | 932708-14-0 | 99.4% | 456.27 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY10918610MM/1ML
Encompass_Preferred
Pexopiprant is an oral antagonist of the prostaglandin D2 receptor 2 (DP2), with a Ki < 100 nM. It is for research use only and not sold to patients.
  • Used in studies of asthma.
  • Oral antagonist of the prostaglandin D2 receptor 2 (DP2).
  • Ki of less than 100 nM.
  • For research use only.
Catalog No. 50-003-45057