Learn More
Medchemexpress LLC PF-00356231 hydrochloride | 820223-77-6 | 99.5% | 464.96 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY114091
Encompass_Preferred
PF-00356231 hydrochloride is a specific, non-peptidic, non-zinc chelating ligand and inhibitor of matrix metalloproteinase MMP-12 (IC50=1.4 μM). It forms a complex with MMP-12 and also demonstrates potency against MMP-13, MMP-8, MMP-9, and MMP-3.
  • Specific inhibitor of MMP-12
  • Non-peptidic, non-zinc chelating ligand
  • Potent against MMP-13, MMP-8, MMP-9, and MMP-3
  • For research use only
Catalog No. 50-004-38229