Learn More
Medchemexpress LLC HY-137048 5mg , PF-00835231 CAS:870153-29-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1370485MG
Encompass_Preferred
HY-137048 5mg PF-00835231 CAS: 870153-29-0 PF-00835231 is a CoV-2 cysteine 3C-like protease (3CLpro) inhibitor, with IC50s of 0.27 nM and 4 nM for SARS CoV-2 and SARS CoV-1 3CLpro, respectively. PF-00835231 is developed for the research of anti-SARS-CoV-2/COVID-19. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9247