Learn More
Medchemexpress LLC PF-00835231 | 870153-29-0 | 99.3% | 472.53 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY137048100MG
Encompass_Preferred
PF-00835231 is a CoV-2 cysteine 3C-like protease (3CLpro) inhibitor, with IC50s of 0.27 nM for SARS CoV-2 and 4 nM for SARS CoV-1 3CLpro. It is developed for the research of anti-SARS-CoV-2/COVID-19 and can inhibit cell infections as well as suppress infections in animal models.
  • Acts as a CoV-2 cysteine 3C-like protease inhibitor.
  • Shows potent inhibition against SARS CoV-2 and SARS CoV-1 3CLpro.
  • Developed for research in anti-SARS-CoV-2/COVID-19.
  • Capable of inhibiting cell infections.
  • Demonstrates suppression of infections in animal models.
  • Effectively inhibits SARS-CoV-2 USA-WA1/2020.
  • Inhibits rhinovirus HRV3C protease.
  • Reduces multifocal lung lesions and lung viral load in SARS-CoV-MA15 infected mouse models.
Catalog No. 50-003-50627