Learn More
Medchemexpress LLC PF-00835231 | 870153-29-0 | 99.31% | 472.53 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13704850MG
Encompass_Preferred
PF-00835231 is a CoV-2 cysteine 3C-like protease (3CLpro) inhibitor with IC50s of 0.27 nM and 4 nM for SARS CoV-2 and SARS CoV-1 3CLpro, respectively. It is developed for the research of anti-SARS-CoV-2/COVID-19, inhibiting cell infections and suppressing infections in animal models. It shows strong inhibitory effects and broad-spectrum activity against coronavirus Mp, with inhibitory effects against SARS-CoV-2 Wuhan, Washington strain 1, and Belgium/GHB-03021/2020 strain infections in various cell lines. It also reduces lung lesions and viral load in infected mouse models.
  • CoV-2 cysteine 3C-like protease inhibitor
  • Inhibits cell infections and suppresses infections in animal models
  • Broad-spectrum activity against coronavirus Mp
  • Effective against SARS-CoV-2 Wuhan, Washington strain 1, and Belgium/GHB-03021/2020 strain infections
  • Reduces lung lesions and viral load in infected mouse models
Catalog No. 50-003-48766