Learn More
Apexbio Technology LLC PF-03084014 865773-15-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A37115.1
Encompass_Preferred
PF-03084014 is a small-molecule inhibitor targeting -secretase It is designed to selectively inhibit -secretase activity thereby blocking the proteolytic cleavage of Notch receptors and regulating the Notch signaling pathway PF-03084014 exerts its biological activity primarily by preventing -secretase-mediated cleavage which inhibits the release of Notch intracellular domain (NICD) fragments and reduces Notch pathway signaling Based on these pharmacological properties PF-03084014 holds research potential in anticancer applications as evaluated in preclinical tumor models such as leukemia and pancreatic adenocarcinoma both as a single agent and in combination strategies
Catalog No. 50-000-95447