Learn More
Cayman Chemical PF-03550096 5mg
SDP
Supplier:  Cayman Chemical 156205 MG
Encompass_Preferred
A synthetic CB that selectively targets the peripheral CB2 receptor over the central CB1 receptor (Kis = 7.9 and 1,500 nM for human isoforms, respectively); intended for forensic and research applications
Catalog No. 50-297-1705