Learn More
TARGETMOL CHEMICALS INC PF-04457845 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T432325MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. PF-04457845 is a greatly and effctive FAAH inhibitor, and for hFAAH(IC50=7.2±0.63 nM) and rFAAH(IC50=7.4±0.62 nM). Purity 100%
Catalog No. 50-287-9930