Learn More
TARGETMOL CHEMICALS INC PF 04531083 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T1651410MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF 04531083 is a selective blocker of the NaV1.8 channel. PF 04531083 can be used for neuropathic and inflammatory pain studies. purity: 99%
Catalog No. 50-258-1172