Learn More
Medchemexpress LLC PF-04745637 | 1917294-46-2 | 100.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HY12068910MM 1ML
Encompass_Preferred
PF-04745637 is a potent and selective TRPA1 antagonist with an IC50 of 17 nM for human TRPA1. It is supplied for research use in solution (10 mM in DMSO, 1 mL) and as multiple solid quantities for laboratory studies. Storage recommendations are provided to preserve stability.
  • Potent TRPA1 inhibition with IC50 17 nM.
  • High purity suitable for research applications.
  • Available as 10 mM solution in DMSO and multiple solid sizes.
  • Recommended storage conditions for powder and solution to maintain stability.
  • Intended for in vitro and topical pharmacology studies.
Catalog No. 50-002-30386