Learn More
TARGETMOL CHEMICALS INC PF-04753299 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T412755MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. PF-04753299 is a potent and selective inhibitor of UDP-3-O-(R-3-hydroxymyristol)-N-acetylglucosamine deacetylase (LpxC). PF-04753299 is bactericidal for the gonococcal isolates. PF-04753299 inhibits E. coli P. aeruginosa and K. pneumoniae strains with MIC 90 values of 2 ug/ml 4 ug/ml and 16 ug/ml respectively. PF-04753299 is used for the study of gram-negative bacteria infection [1]. purity: 99%
Catalog No. 50-257-9942