Learn More
Medchemexpress LLC HY-122026 5mg , PF-04802367 CAS:1962178-27-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1220265MG
Encompass_Preferred
HY-122026 5mg PF-04802367 CAS: 1962178-27-3 PF-04802367 (PF-367) is a highly selective GSK-3 inhibitor with an IC50 of 2.1 nM based on a recombinant human GSK-3β enzyme assay and 1.1 nM based on ADP-Glo assay. PF-04802367 shows desirable central nervous system (CNS) properties and potency. PF-04802367 is equally effective at inhibition of the two known GSK-3 isoforms (GSK-3α and GSK-3β) with IC50 values of 10.0 and 9.0 nM in mobility shift assays, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1321