Learn More
Medchemexpress LLC PF-04859989 hydrochl 10mM 1mL | 177943-33-8 | 214.65 g/mol | C9H11ClN2O2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11645110MM 1ML
Encompass_Preferred
PF-04859989 hydrochloride is a brain-penetrant, irreversible kynurenine aminotransferase II (KAT II) inhibitor used in research to probe KAT II-mediated pathways. It shows low-nanomolar potency against human KAT II and is supplied as a concentrated solution for in vitro biochemical and cellular assays.
  • Brain-penetrant irreversible KAT II inhibitor suitable for research use.
  • Reported IC50 ~23 nM for human KAT II, indicating high potency.
  • Supplied as a 10 mM solution in DMSO for direct preparation of assay dilutions.
  • High chemical purity appropriate for biochemical and cellular studies.
  • Certificate of analysis and safety data sheet available to support experimental use.
Catalog No. 50-002-26181