Learn More
TARGETMOL CHEMICALS INC PF-04929113 MESYLATE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T43425MG
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PF-04929113 Mesylate (SNX-5422 Mesylate) a prodrug of SNX-2112 is an orally available Hsp90 inhibitor (Kd 41 nM) and also induces Her-2 degradation (IC50 37 nM). purity: 99%
Catalog No. 50-257-9905